missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Alanyl-L-glutamine, 97%

Product Code. 15264874 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15264874 5 g Glass bottle 5g
15244874 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15264874 Supplier Thermo Scientific Chemicals Supplier No. 451610050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39537-23-0 | C8H15N3O4 | 217.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39537-23-0
Molecular Formula C8H15N3O4
Molecular Weight (g/mol) 217.23
MDL Number MFCD00133046
InChI Key HJCMDXDYPOUFDY-WHFBIAKZSA-N
Synonym l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax
PubChem CID 123935
ChEBI CHEBI:73788
IUPAC Name (2S)-5-amino-2-[[(2S)-2-aminopropanoyl]amino]-5-oxopentanoic acid
SMILES C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 213.0°C to 220.0°C
Ignition Residue 0.1% max
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 217.22
Percent Purity 97%
Chemical Name or Material L-Alanyl-L-glutamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.