missing translation for 'onlineSavingsMsg'
Learn More

L-allo-Isoleucine, 99%, 99% ee

Product Code. p-6827177 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10081024 100 mg 100mg
10113294 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081024 Supplier Thermo Scientific Acros Supplier No. 399491000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1509-34-8 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1509-34-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00066446
InChI Key AGPKZVBTJJNPAG-BJNJOQLTNA-N
Synonym l-allo-isoleucine, l-alloisoleucine, alloisoleucine, 2s,3r-2-amino-3-methylpentanoic acid, allo-isoleucine, allo-l-isoleucine, h-allo-ile-oh, 3r-ls-isoleucine, isoleucine, allo, isoleucine, threo
PubChem CID 99288
ChEBI CHEBI:43433
IUPAC Name (2S,3R)-2-amino-3-methylpentanoic acid
SMILES CC[C@@H](C)[C@H](N)C(O)=O

Specifications

Assay Percent Range 99%
Linear Formula CH3CH2CH(CH3)CH(NH2)COOH
Quantity 100 mg
Formula Weight 131.17
Chemical Name or Material L-allo-Isoleucine, 99%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.