missing translation for 'onlineSavingsMsg'
Learn More

L-alpha-Dipalmitoyl phosphatidylcholine, 98%

Product Code. 10626974 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Unit Size:
100mg
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10626974 100 mg 100mg
10134670 500 mg 500mg
10512174 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10626974 Supplier Thermo Scientific Acros Supplier No. 240571000
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63-89-8 | C40H80NO8P | 734.053 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-89-8
Molecular Formula C40H80NO8P
Molecular Weight (g/mol) 734.053
MDL Number MFCD00036903
InChI Key KILNVBDSWZSGLL-KXQOOQHDSA-N
Synonym colfosceril palmitate, dppc, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine, 1,2-dipalmitoyl-l-lecithin, dipalmitoylphosphatidylcholine, 1,2-dihexadecanoyl-sn-glycero-3-phosphocholine, colfoscerili palmitas, palmitate de colfosceril, palmitato de colfoscerilo, unii-319x2nfw0a
PubChem CID 452110
ChEBI CHEBI:72999
IUPAC Name [(2R)-2,3-di(hexadecanoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCC

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 mg
Specific Rotation Condition + 7.00 (20.00°C c=2,CHCl3)
Specific Rotation +6.0° to +8.0° (20°C, 589nm) (c=2, CHCl3)
Formula Weight 734.08
Percent Purity ≥97.5% (Fatty acid purity)
Physical Form Powder
Chemical Name or Material L-α-Dipalmitoyl phosphatidylcholine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.