missing translation for 'onlineSavingsMsg'
Learn More

L-Anserine, TRC

Product Code. 30012728
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012728 5 g 5g
30010609 10 g 10g
30006699 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012728 Supplier Toronto Research Chemicals Supplier No. TRCA6782535G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-85-0
Molecular Formula C10 H16 N4 O3
Molecular Weight (g/mol) 240.26
Synonym β-Alanyl-3-methyl-L-histidine, Anserine (8CI), Histidine, N-β-alanyl-3-methyl-, L- (7CI), L-Histidine, N-β-alanyl-3-methyl-, β-Alanyl-3-methyl-L-histidine, 1: PN: WO2009033771 PAGE: 100 claimed protein, L-Anserine, Marine Active
IUPAC Name (2S)-2-(3-aminopropanoylamino)-3-(3-methylimidazol-4-yl)propanoic acid
SMILES Cn1cncc1C[C@H](NC(=O)CCN)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H16N4O3/c1-14-6-12-5-7(14)4-8(10(16)17)13-9(15)2-3-11/h5-6,8H,2-4,11H2,1H3,(H,13,15)(H,16,17)/t8-/m0/s1
Formula Weight 240.1222
Percent Purity >95%
Chemical Name or Material L-Anserine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.