missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine-13C6,15N4 Hydrochloride, TRC

Product Code. 30136295
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136295 25 mg 25mg
30008720 10 mg 10mg
30004634 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136295 Supplier Toronto Research Chemicals Supplier No. TRCA76950425MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202468-25-5
Molecular Formula 13C6 H14 15N4 O2 . H Cl
Molecular Weight (g/mol) 220.59
Synonym L-Arginine-guanidino,1,2,3,4,5-13C6-guanidino,guanidino,guanidino,N2-15N4, hydrochloride (1:1), L-Arginine-13C6-15N4, monohydrochloride (9CI), 13C615N4 L-Arginine-hydrochloride, L-Arginine-guanidino,1,2,3,4,5-13C6-guanidino,guanidino,guanidino,N2-15N4 hydrochloride, L-Arginine-13C6,15N4 hydrochloride, (2S)-2-(Amino-15N)-5-[(diaminomethylidene-13C,15N2)amino-15N]pentanoic-1,2,3,4,5-13C5 acid hydrochloride, (2S)-2-Amino-5-carbamimidamidopentanoic-13C6,15N4 acid hydrochloride, (S)-2-(Amino-15N)-5-(guanidino-13C,15N3)pentanoic-1,2,3,4,5-13C5 acid hydrochloride, (S)-Arginine-13C6,15N4 hydrochloride
IUPAC Name (2S)-2-(15N)azanyl-5-[(15N-azanyl-13C-carbon-15N-imidoyl)15N-amino](1,2,3,4,5-13C5)pentanoic acid;hydrochloride
SMILES Cl.[15NH2][13C@@H]([13CH2][13CH2][13CH2][15NH][13C](=[15NH])[15NH2])[13C](=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1;
Formula Weight 220.0966
Chemical Name or Material L-Arginine hydrochloride 13C6, 15N4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.