missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, 99%, MP Biomedicals™

Product Code. 11323929 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323929 1 kg 1kg
11303929 100 g 100g
11313929 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323929 Supplier MP Biomedicals Supplier No. 0210073691
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74-79-3 | C6H14N4O2 | 174.20 g/mol

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Color White
Assay Percent Range 99%
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
Quantity 1 kg
Formula Weight 174.2
Grade Cell Culture
Physical Form Powder
Chemical Name or Material L-Arginine free base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.