missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, Free Base, Fisher BioReagents

Product Code. 10041693
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041693 100 g Poly Bottle 100g
10421725 500 g Poly Bottle 500g
10368364 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10041693 Supplier Fisher BioReagents Supplier No. BP2505100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
L-Arginine is an essential amino acid used in biochemical research applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Melting Point 223°C
pH 11.4
Color White
Identification Pass Test
Infrared Spectrum Conforms to reference
Assay Percent Range ≥99 %
Loss on Drying 0.3% max.
Packaging Poly Bottle
Specific Rotation +26.3 to +27.7° (+ or -)
Merck Index 15, 770
Quantity 100 g
Formula Weight 174.12
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material L-Arginine, Free Base
Show More Show Less
compliance-icons
Nombre del producto
  • L(+)-Arginine
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
Indicaciones de peligro
  • H319-Causes serious eye irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.