missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine hydrochloride, 98.5 to 101.5%

Product Code. 15855258 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15855258 1 kg Plastic Bottle 1kg
15875088 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15855258 Supplier Thermo Scientific Supplier No. J11500.A1
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1119-34-2 | C6H15ClN4O2 | 210.662 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
MDL Number MFCD00064550
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2 S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Melting Point 216°C (decomposition)
Packaging Plastic Bottle
Linear Formula NH2C(NH)NH(CH2)3CH(NH2)COOH.HCl
Beilstein 3631658
Density 1.42
Quantity 1 kg
Formula Weight 210.66
Percent Purity 98.5 to 101.5%
Specific Gravity 1.42
Physical Form Crystalline Powder
Chemical Name or Material L-Arginine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.