missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Arginine Hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 10772564 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10772564

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H14N4O2·HCl
Molecular Weight (g/mol) 210.67
MDL Number MFCD00064550
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Melting Point 226°C
Color White
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
Quantity 500 g
Merck Index 15,77
Specific Rotation Condition +22° (20°C c=8,6N HCl)
Specific Rotation + 22
Solubility Information Solubility in water: 75.1g/100g (20°C)
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Formula Weight 210.67
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Arginine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.