missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Arginine hydrochloride, MedChemExpress
Description
L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) is the nitrogen donor for synthesis of nitric oxide, a potent vasodilator that is deficient during times of sickle cell crisis.
Specifications
Specifications
| Chemical Name or Material | L-Arginine hydrochloride |
| CAS | 1119-34-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C6H15ClN4O2 |
| Synonym | (S)-(+)-Arginine hydrochloride |
| Solubility Information | H2O : 100 mg/mL (474.70 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (4.75 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | N[C@@H](CCCNC(N)=N)C(O)=O.[H]Cl |
| Molecular Weight (g/mol) | 210.66 |
| Quantity | 5 g |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?