missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, Hydrochloride, MP Biomedicals™

Product Code. 11373929 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373929 100 g 100g
11383929 500 g 500g
11393929 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373929 Supplier MP Biomedicals Supplier No. 0210074380
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

MDL Number MFCD00002635
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467

Specifications

CAS 1119-34-2
Color White
Assay Percent Range 0.992
Molecular Formula C6H14N4O2
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
SMILES NC(CCCN=C(N)N)C(O)=O
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Weight (g/mol) 174.20
Quantity 100 g
Formula Weight 210.662
Physical Form Crystalline Powder
Chemical Name or Material L-Arginine Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.