missing translation for 'onlineSavingsMsg'
Learn More

L-arginine methyl ester dihydrochloride, 98%, ACROS Organics™

Product Code. 10240133 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10240133 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10240133 Supplier Thermo Scientific Acros Supplier No. 376680050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 26340-89-6
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C7H18Cl2N4O2
Packaging Glass Bottle
Linear Formula NH2C(NH)N(H)(CH2)3CH(NH2)C(O)OCH3·2HCl
MDL Number MFCD00038948
Specific Rotation +18° to +22° (20°C, 589nm) (c=2.5, CH3OH)
Synonym l-arginine methyl ester dihydrochloride, h-arg-ome.2hcl, methyl l-argininate dihydrochloride, h-arg-ome 2hcl, l-arginine, methyl ester, dihydrochloride, s-methyl 2-amino-5-guanidinopentanoate dihydrochloride, methyl 2s-2-amino-5-carbamimidamidopentanoate dihydrochloride, l-arginine, methyl ester, hydrochloride 1:2, h-arg-ome?hcl, h-arg-ome 2 hcl
InChI Key XQYZOBNLCUAXLF-XRIGFGBMSA-N
SMILES COC(=O)C(CCCN=C(N)N)N.Cl.Cl
IUPAC Name methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
Specific Rotation Condition +20° (20°C c=2.5 Chloroform)
Molecular Weight (g/mol) 261.147
Quantity 5 g
PubChem CID 117776
Formula Weight 261.15
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Arginine methyl ester dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.