missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10143483
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
10143483 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 10143483

Brand: Fisher BioReagents BP370100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 223°C
Ammonium 0.02% max.
CAS 74-79-3
Arsenic (As) 1ppm max.
pH 11.4
Color White
Heavy Metals (as Pb) 10ppm max.
Identification Pass Test
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.5% max.
Other Amino Acids Not Detected
Molecular Formula C6H14N4O2
Packaging Glass Bottle
MDL Number MFCD00002635
Residue after Ignition 0.1% max.
Specific Rotation 26.9 to 27.9° (+ or -)
Merck Index 15, 770
State of Solution (%T) 98% min.
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
Alpha Vector Arginine
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
SMILES NC(CCCN=C(N)N)C(O)=O
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
Molecular Weight (g/mol) 174.20
Quantity 100 g
PubChem CID 6322
ChEBI CHEBI:16467
Formula Weight 174.2
Percent Purity 98.5 to 101.0%
Physical Form Solid
Chemical Name or Material L-Arginine
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Arginine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.