missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic acid 2-phosphate trisodium, MedChemExpress

Product Code. 16471089
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471089 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471089 Supplier Medchem Express Supplier No. HY107837
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Ascorbic acid 2-phosphate trisodium (2-Phospho-L-ascorbic acid trisodium) is a long-acting vitamin C derivative that can stimulate collagen formation and expression.

L-Ascorbic acid 2-phosphate trisodium (2-Phospho-L-ascorbic acid trisodium) can be used as a culture medium supplement for the osteogenic differentiation of human adipose stem cells (hASCs). L-Ascorbic acid 2-phosphate trisodium (2-Phospho-L-ascorbic acid trisodium) increases alkaline phosphatase (ALP) activity and expression of runx2A in hASCs during the osteogenic differentiation.

Specifications

Chemical Name or Material L-Ascorbic acid 2-phosphate trisodium
CAS 66170-10-3
Color White
Purity Grade Notes Research
Molecular Formula C6H6Na3O9P
Quantity 1 g
Synonym 2-Phospho-L-ascorbic acid trisodium
Solubility Information H2O : 150 mg/mL (465.77 mM; Need ultrasonic) ∣DMSO : 6 mg/mL (18.63 mM; Need ultrasonic and warming)
SMILES O[C@@H](CO)[C@@H](O1)C(O[Na])=C(OP(O[Na])(O[Na])=O)C1=O
Molecular Weight (g/mol) 322.05
Formula Weight 322.05
Percent Purity 98.57%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.