missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ascorbic acid, ACS reagent

Product Code. 10480831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10480831 500 g Plastic bottle 500g
10055800 100 g Plastic bottle 100g
10712354 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10480831 Supplier Thermo Scientific Acros Supplier No. 401475000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-81-7 | C6H8O6 | 176.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point 190.0°C to 192.0°C
Color White to Yellow
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 500 g
Beilstein 18, IV, 3038
Merck Index 15, 819
Specific Rotation + 20.50
Solubility Information Solubility in water: 333g/L (20°C). Other solubilities: ethanol 20g/L (20°C), soluble methanol and acetone, insoluble in ether, chloroform and benzene
Formula Weight 176.13
Percent Purity ≥99.0%
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material L(+)-Ascorbic acid, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.