missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid, Certified AR for Analysis, Fisher Chemical

Product Code. 10713791 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10713791 100 g 100g
10559950 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713791 Supplier Fisher Chemical Supplier No. A/8882/48
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C6H8O6, CAS Number-50-81-7, 500g, White, 176.13g/mol, 190 deg.C, 64328, Plastic powder jar, 2.1 to 2.6, Solid | CAS: 50-81-7 | C6H8O6

C6H8O6, CAS Number-50-81-7, 500g, White, 176.13g/mol, 190 deg.C, 64328, Plastic powder jar, 2.1 to 2.6, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
MDL Number 64328

Specifications

Melting Point 190°C
Color White
pH 2.1 to 2.6
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 176.13g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.