missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid Crystalline MP Biomedicals

Product Code. 11373949 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373949 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11373949 Supplier MP Biomedicals Supplier No. 0210076991
 Restricted Item
Estimated Shipment: 23-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 50-81-7
Density 1.65 (water = 1)
Molecular Formula C6H8O6
MDL Number MFCD00064328
Quantity 1 kg
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES C(C(C1C(=C(C(=O)O1)O)O)O)O
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Molecular Weight (g/mol) 176.12
PubChem CID 54670067
ChEBI CHEBI:29073
Formula Weight 176.12
Physical Form Crystalline
Chemical Name or Material L-Ascorbic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.