missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic acid sodium salt, 99%

Product Code. 11443358
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443358 500 g 500g
11433358 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443358 Supplier Thermo Scientific Alfa Aesar Supplier No. A17759.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 134-03-2 | C6H10NaO6 | 201.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
L-Ascorbic acid sodium salt is used as an antioxidant, an acidity regulator and an antimicrobial in food stuffs. As a reductant, it is involved in the isolation of chloroplasts. It acts as a coenzyme for many hydroxylation reactions. It plays a vital role to eliminate chronic and acute infections. It finds applications as an additive in curing meat products such as ham and sausage meat. It is widely used as a nutritional supplement in agriculture and animal feed to promote the growth of young animals and prevent scurvy. It is also used as an intermediate in the manufacture of various organic chemicals.

Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in chloroform and ether.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00082340
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
PubChem CID 131674100
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point ∼220°C
Quantity 500 g
Beilstein 3767246
Merck Index 14,830
Solubility Information Soluble in water. Slightly soluble in alcohol. Insoluble in chloroform and ether.
Optical Rotation +105° (c=5 in Water)
Formula Weight 198.11
Percent Purity 99%
Chemical Name or Material L-Ascorbic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.