missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid Sodium Salt, MP Biomedicals

Product Code. 11446272 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
Unit Size:
1kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446272 1 kg 1kg
11426272 25 g 25g
11436272 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446272 Supplier MP Biomedicals Supplier No. 0210289091
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00082340
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
PubChem CID 131674100
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 1 kg
Formula Weight 198.11
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material L-Ascorbic Acid Sodium Salt, Antimicrobial, Antioxidant, Reductant for the isolation of chloroplasts
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.