missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid, TRC

Product Code. 17947864
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17947864 5 g 5g
17919114 1 g 1g
17959944 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17947864 Supplier Toronto Research Chemicals Supplier No. TRCA7869905G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6 H8 O6
Molecular Weight (g/mol) 176.12
Synonym (+)-Ascorbic acid, 100M, 3-Oxo-L-gulofuranolactone, 3-keto-L-Gulofuranolactone, 5-(1,2-Dihydroxyethyl)-3,4-dihydroxy-2(5H)-furanone, 5-(1,2-Dihydroxyethyl)-3,4-dihydroxy-5H-furan-2-one, 5-(1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Adenex, Allercorb, Antiscorbic vitamin, Antiscorbutic vitamin, Ascoltin, Ascorbajen, Ascorbalox, Ascorbic acid, Ascorbicap, Ascorbutina, Ascorell, Ascorin, Ascorteal, Ascorvit, Bovi-C, C-L 6/PW, C-Quin, C-Vimin, Cantan, Cantaxin, Catavin C, Ce-Mi-Lin, Ce-Vi-Sol, Cebicure, Cebion, Cebione, Cecon, Cegiolan, Ceglion, Ceklin, Celaskon, Celin, Cell C, Cemagyl, Cenetone, Cereon, Cergona, Cescorbat, Cetamid, Cetane, Cetane-Caps TC, Cetebe, Cetemican, Cevalin, Cevarol, Cevatine, Cevex, Cevimin, Cevital, Cevitamic acid, Cevitamin, Cevitan, Cevitex, Cewin, Chewcee, Ciamin, Cinal, Cipca, Citrovit, Colascor, Concemin, Davitamon C, E 300, Emvit-C, Energil C, Hicee, Hybrin, IDO-C, Juvamine, Kangbingfeng, L-(+)-Ascorbic acid, L-3-Keto-threo-hexuronic acid lactone, L-Lyxoascorbic acid, L-Xyloascorbic acid, L-threo-Ascorbic acid, L-threo-Hex-2-enonic acid, γ-lactone, Laroscorbine, Lemascorb, Liqui-Cee, Monovitan C, NSC 218455, NSC 33832, Neo-Valdrin, Novasol Cof, P 1110, Pascorbin, Planavit C, Proscorbin, Redoxon, Ribena, Ronotec 100, Rontex 100, Rovimix C, Rovimix C-EC, Scorbu C, Secorbate, Suncoat VC 40, Tanvimil C, Testascorbic, VC 40, VC 97, Vaginorm-C, Vasc, Vicelat, Vicin, Viforcit, Viscorin, Viscorin 100M, Vitace, Vitacee, Vitacimin, Vitacin, Vitamin C, Vitamisin, Vitascorbol, Vitashure 160, Xitix, Xyloascorbic acid, L-
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(=C1O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1
Formula Weight 176.0321
Percent Purity >95%
Chemical Name or Material L-Ascorbic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.