missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals

Product Code. 15100337 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15100337 25 g 25g
15214774 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15100337 Supplier Thermo Scientific Acros Supplier No. 449480250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 137-66-6 | Molecular Formula: C22H38O7 | Molecular Weight: 414.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-66-6
Molecular Formula C22H38O7
Molecular Weight (g/mol) 414.54
MDL Number MFCD00005377
InChI Key NULLRXHRYOXSEW-UHFFFAOYNA-N
Synonym ascorbyl palmitate, l-ascorbyl 6-palmitate, 6-o-palmitoyl-l-ascorbic acid, ascorbic palmitate, l-ascorbyl palmitate, unii-qn83us2b0n, ascorbyl monopalmitate, l-ascorbic acid 6-palmitate, s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate, ascorbyl palmitate nf
PubChem CID 54680660
IUPAC Name [(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O

Specifications

Melting Point 111°C to 115°C
Boiling Point 250°C
Ignition Residue 0.1% max
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 414.53
Chemical Name or Material L(+)-Ascorbyl palmitate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.