missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine, 99%

Product Code. 10564654 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10564654

Brand: Thermo Scientific Acros 371600025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00064401
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic Bottle
Linear Formula H2NCOCH2CH(NH2)CO2H
Specific Rotation + 34.50
Solubility Information Solubility in water: 20g/l (20°C). Other solubilities: insoluble in organic solvents
Water 1% max.
Specific Rotation Condition + 34.50 (20.00°C c=10,2N HCl)
Quantity 2.5 kg
Formula Weight 132.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-Asparagine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.