missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine, MedChemExpress

Product Code. 16419126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419126 Supplier Medchem Express Supplier No. HYN0667100mg
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Asparagine ((-)-Asparagine) is a non-essential amino acid that is involved in the metabolic control of cell functions in nerve and brain tissue.

L-Asparagine ((-)-Asparagine) is a non-essential amino acid that is involved in the metabolic control of cell functions in nerve and brain tissue.

Specifications

Chemical Name or Material L-Asparagine
CAS 70-47-3
Color White
Purity Grade Notes Research
Molecular Formula C4H8N2O3
Synonym (-)-Asparagine Asn Asparamide
Solubility Information H2O : 6.67 mg/mL (50.48 mM; Need ultrasonic)
SMILES N[C@H](C(O)=O)CC(N)=O
Molecular Weight (g/mol) 132.12
Quantity 100 mg
Formula Weight 132.12
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.