missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartate de magnesium salt, MP Biomedicals™

Product Code. 11493251 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
Unit Size:
1kg
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493251 1 kg 1kg
11483251 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493251 Supplier MP Biomedicals Supplier No. 0215040691
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity magnesium salt of L-aspartic acid for research or manufacturing applications.

  • CAS Number: 18962-61-3
  • Molecular Formula: C8H12N2O8Mg
  • Molecular Weight: 288.5 (anhydrous)
  • EC Number: 242-703-7
  • MDL Number: MFCD00044935
  • Optical Rotation: +20°
  • pH: 6.5
  • Solubility: Freely soluble in water
  • Hazard Statement: H317
  • Usage: For research or further manufacturing use only; not for direct human use

Chemical Identifiers

CAS 18962-61-3
Molecular Formula C8H10MgN2O8-2
Molecular Weight (g/mol) 286.479
InChI Key XBYSRGUYNYEVEJ-UHFFFAOYSA-L
Synonym l-aspartic acid magnesium salt
PubChem CID 131850428
IUPAC Name magnesium;2-azanidylbutanedioate;hydron
SMILES [H+].[H+].C(C(C(=O)[O-])[NH-])C(=O)[O-].C(C(C(=O)[O-])[NH-])C(=O)[O-].[Mg+2]

Specifications

Color White
Quantity 1 kg
Formula Weight 288.5 (Anhydrous)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartate de magnesium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.