missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid 4-tert-butyl ester, 98%

Product Code. 15446228
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446228 1 g 1g
15456228 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446228 Supplier Thermo Scientific Alfa Aesar Supplier No. H59810.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3057-74-7 | C8H15NO4 | 189.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3057-74-7
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
MDL Number MFCD00038577
InChI Key MXWMFBYWXMXRPD-YFKPBYRVSA-N
Synonym h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh
PubChem CID 7010570
IUPAC Name (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CC(C(=O)O)N

Specifications

Beilstein 4671089
Quantity 1 g
Formula Weight 189.21
Optical Rotation −9° (c=2 in Methanol: Water (4:1))
Percent Purity 98%
Chemical Name or Material L-Aspartic acid 4-tert-butyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.