missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Aspartic Acid Dibenzyl Ester p-Toluenesulfonate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | L-Aspartic Acid Dibenzyl Ester p-Toluenesulfonate Salt |
| CAS | 2886-33-1 |
| Molecular Formula | C25H27O7S |
| Quantity | 5 g |
| Synonym | L-Aspartic Acid 1,4-bis(Phenylmethyl) Ester 4-Methylbenzenesulfonate, Aspartic Acid Dibenzyl Ester p-Toluenesulfonate, L-Aspartic Acid bis(Phenylmethyl) Ester 4-Methylbenzenesulfonate, Dibenzyl L-Aspartate Tosylate, Dibenzyl Aspartate p-Tosylate, Dibenzyl Aspartate Tosylate, Dibenzyl Aspartate Tosylate Salt, L-Aspartic Acid Dibenzyl Ester 4-Toluenesulfonate Salt, L-Aspartic Acid Dibenzyl Ester p-toluenesulfonate, L-Aspartic Acid Dibenzyl Ester p-Toluenesulfonate Salt, L-Aspartic Acid Dibenzyl Ester Tosylate, L-Aspartic Acid alpha,alpha-Dibenzyl Ester Tosylate, S-Aspartic Acid Dibenzyl Ester p-Tosylate |
| InChI Formula | InChI=1S/C18H19NO4.C7H8O3S/c19-16(18(21)23-13-15-9-5-2-6-10-15)11-17(20)22-12-14-7-3-1-4-8-14;1-6-2-4-7(5-3-6)11(8,9)10/h1-10,16H,11-13,19H2;2-5H,1H3,(H,8,9,10)/t16-;/m0./s1 |
| SMILES | Cc1ccc(cc1)S(=O)(=O)O.N[C@@H](CC(=O)OCc2ccccc2)C(=O)OCc3ccccc3 |
| IUPAC Name | dibenzyl (2S)-2-aminobutanedioate;4-methylbenzenesulfonic acid |
| Molecular Weight (g/mol) | 485.55 |
| Formula Weight | 485.1508 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?