missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid diethyl ester hydrochloride, 98%

Product Code. 15480969
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480969 25 g 25g
15470969 5 g 5g
15418616 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480969 Supplier Thermo Scientific Alfa Aesar Supplier No. H63974.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16115-68-7 | C8H16ClNO4 | 225.669 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16115-68-7
Molecular Formula C8H16ClNO4
Molecular Weight (g/mol) 225.669
MDL Number MFCD00038910
InChI Key AJOXZAAREAYBQR-RGMNGODLSA-N
Synonym l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride
PubChem CID 12416466
IUPAC Name diethyl (2S)-2-aminobutanedioate;hydrochloride
SMILES CCOC(=O)CC(C(=O)OCC)N.Cl

Specifications

Quantity 25 g
Formula Weight 225.67
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Aspartic acid diethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.