missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid dimethyl ester hydrochloride, 98%

Product Code. 10371763 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10371763 5 g Glass Bottle 5g
10563143 25 g Plastic Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10371763 Supplier Thermo Scientific Supplier No. 376660050
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32213-95-9 | C6H12ClNO4 | 197.615 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32213-95-9
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.615
MDL Number MFCD00038878
InChI Key PNLXWGDXZOYUKB-WCCKRBBISA-N
Synonym h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
PubChem CID 2734892
IUPAC Name dimethyl (2S)-2-aminobutanedioate;hydrochloride
SMILES COC(=O)CC(C(=O)OC)N.Cl

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3OC(O)CH2CH(NH2)C(O)OCH3·HCl
Specific Rotation +13° to +17° (20°C, 589nm) (c=1, CH3OH)
Specific Rotation Condition +15° (20°C c=1,Chloroform)
Quantity 5 g
Formula Weight 197.62
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartic acid dimethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.