missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid monosodium salt monohydrate, 97%

Product Code. 15284874 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15284874 25 g Plastic Bottle 25g
15204884 100 g Plastic Bottle 100g
15274874 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15284874 Supplier Thermo Scientific Acros Supplier No. 451620250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3792-50-5 | C4H11NNaO6 | 192.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3792-50-5
Molecular Formula C4H11NNaO6
Molecular Weight (g/mol) 192.123
InChI Key VJDUMAWJHIHDHJ-SQGDDOFFSA-N
Synonym monosodium l-aspartate dihydrate
PubChem CID 129893661
IUPAC Name (2S)-2-aminobutanedioic acid;sodium;dihydrate
SMILES C(C(C(=O)O)N)C(=O)O.O.O.[Na]

Specifications

Assay Percent Range 97%
Packaging Plastic Bottle
Quantity 25 g
Formula Weight 173.1
Percent Purity 97%
Chemical Name or Material L-Aspartic acid monosodium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.