missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartic acid monosodium salt monohydrate, 99%

Product Code. 11486016
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486016 500 g 500g
11476016 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486016 Supplier Thermo Scientific Chemicals Supplier No. B22321.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 323194-76-9 | C4H8NNaO5 | 173.10 g/mol

L-Aspartic acid monosodium salt monohydrate is used as a principal neurotransmitter for fast synaptic excitation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid monosodium salt monohydrate is used as a principal neurotransmitter for fast synaptic excitation.

Solubility
Soluble in cold water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 323194-76-9
Molecular Formula C4H8NNaO5
Molecular Weight (g/mol) 173.10
MDL Number MFCD00152960
InChI Key PPTHNBYUFXSJPS-UHFFFAOYNA-M
Synonym sodium l-aspartate, l-aspartic acid sodium salt monohydrate, sodium aspartate monohydrate, sodium l-aspartate monohydrate, s-2-aminobutanedioic acid sodium salt, l-aspartic acid monosodium salt monohydrate, l-aspartic acid, monosodium salt, monohydrate, sodium +-aspartate hydrate, l-aspartic acid sodium salt monohydrate tlc, l-aspartic acid sodium salt monohydrate nt
PubChem CID 23679051
SMILES O.[Na+].NC(CC(O)=O)C([O-])=O

Specifications

Solubility Information Soluble in cold water.
IUPAC Name sodium;(2S)-2-amino-4-hydroxy-4-oxobutanoate;hydrate
Quantity 500 g
Formula Weight 173.11 (155.10 Anhydrous)
Percent Purity 99%
Chemical Name or Material L-Aspartic acid monosodium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.