missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid, MP Biomedicals

Product Code. 11343999 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343999 1 kg 1kg
11353999 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343999 Supplier MP Biomedicals Supplier No. 0210080901
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

  • L-Aspartic acid is an endogenous NMDA receptor agonist
  • Principal neurotransmitter for fast synaptic excitation

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-84-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00002616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
PubChem CID 5960
ChEBI CHEBI:17053
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Melting Point 270°C to 271°C (literature)
Color White
Linear Formula C4H7NO4
Solubility Information More soluble in salt solutions; soluble in water acids, alkalies; insoluble in alcohol.
Quantity 1 kg
Formula Weight 133.1
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material L-ASPARTIC ACID
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.