missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid Sodium Salt Monohydrate, ≥98% (dry basis), Thermo Scientific™

Product Code. 15885258 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
10 kg
Packaging:
Plastic bottle
Unit Size:
10kg
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15885258 10 kg Plastic bottle 10kg
15875258 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15885258 Supplier Thermo Scientific Supplier No. J11635.A9
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 323194-76-9
Molecular Formula C4H8NNaO5
Molecular Weight (g/mol) 173.10
MDL Number MFCD00152960
InChI Key PPTHNBYUFXSJPS-UHFFFAOYNA-M
Synonym sodium l-aspartate, l-aspartic acid sodium salt monohydrate, sodium aspartate monohydrate, sodium l-aspartate monohydrate, s-2-aminobutanedioic acid sodium salt, l-aspartic acid monosodium salt monohydrate, l-aspartic acid, monosodium salt, monohydrate, sodium +-aspartate hydrate, l-aspartic acid sodium salt monohydrate tlc, l-aspartic acid sodium salt monohydrate nt
PubChem CID 23679051
IUPAC Name sodium;(2S)-2-amino-4-hydroxy-4-oxobutanoate;hydrate
SMILES O.[Na+].NC(CC(O)=O)C([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 10 kg
Linear Formula NH2CH(COOH)CH2COONa.H2O
Formula Weight 173.1
Percent Purity ≥98%
Packaging Plastic bottle
Chemical Name or Material L-Aspartic acid sodium salt monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.