missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartyl-L-phenylalanine methyl ester, 98%

Product Code. 10093140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10093140 1 g Glass Bottle 1g
10217603 5 g Glass Bottle 5g
10227603 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10093140 Supplier Thermo Scientific Chemicals Supplier No. 228650010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22839-47-0 | C14H18N2O5 | 294.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
Synonym aspartame, nutrasweet, asp-phe-ome, asp-phe methyl ester, aspartam, aspartamum, aspartamo, l-aspartyl-l-phenylalanine methyl ester, aspartylphenylalanine methyl ester, dipeptide sweetener
PubChem CID 134601
ChEBI CHEBI:2877
IUPAC Name (3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O

Specifications

Melting Point 248.0°C to 250.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Loss on Drying 4.5% max. (105°C, 4 hrs)
Packaging Glass Bottle
Linear Formula HO2CCH2CH(NH2)CONHCH(CH2C6H5)CO2CH3
Specific Rotation + 12.50
Merck Index 15, 829
Transmittance 95% min. (c=1, 2N HCl, 430nm) 1cm cell
Solubility Information Solubility in water: sparingly soluble.
Quantity 1 g
Formula Weight 294.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartyl-L-phenylalanine methyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.