missing translation for 'onlineSavingsMsg'
Learn More

L-beta-Proline, 98+%

Product Code. 15454288
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15454288

Brand: Thermo Scientific Alfa Aesar H57895.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72580-53-1 | C5H9NO2 | 115.132 g/mol

It is an agrochemical and pharmacetical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical and pharmacetical intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 8°C. Incompatible with strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72580-53-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD06738463
InChI Key JAEIBKXSIXOLOL-BYPYZUCNSA-N
Synonym s-pyrrolidine-3-carboxylic acid, s-+-pyrrolidine-3-carboxylic acid, l-beta-proline, 3s-3-pyrrolidinecarboxylic acid, 3s-pyrrolidine-3-carboxylic acid, 3-pyrrolidinecarboxylic acid, 3s, s-pyrrolidine-3-carboxylicacid, s-beta-proline, s-, a-proline, pubchem11126
PubChem CID 1501969
IUPAC Name (3S)-pyrrolidine-3-carboxylic acid
SMILES C1CNCC1C(=O)O

Specifications

Melting Point 185°C to 191°C
Beilstein 3537293
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 115.13
Optical Rotation +18° (c=2 in Water)
Percent Purity ≥98%
Chemical Name or Material L-beta-Proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato