missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Nutritionally rich medium originally developed for growth and maintenance of recombinant E. coli strains.
- Supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesize.
- Contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
- Key applications.
- Cell culture
- Proteomics
- Purification and detection
- Recombinant protein expression and analysis
- Bacterial culture
- Plasmid production
- Protein expression
- Gene expression
- Cloning
- Genetic manipulation
- Research Use Only (RUO).
Specifications
Specifications
| Appearance | Powder |
| Certifications/Compliance | Molecular Biology Certified™ |
| Content And Storage | Store dry L-Broth Agar at ambient temperature (15-30°C). Powder medium is hygroscopic; tightly cap; minimize exposure to air. Store reconstituted media at 4°C |
| Form | Powder |
| Grade | Molecular Biology Certified™ Bacterial Growth Medium |
| Media Type | Bacterial media |
| Formulation | Typically contains 10 g Tryptone; 5 g Yeast Extract; 0.5 g NaCl; 15 g Agar. Small quantities of biological buffers have been added for proper pH adjustment. |
| For Use With (Application) | Cell culture, proteomics, purification and detection, recombinant protein expression and analysis, bacterial culture, plasmid production, protein expression, gene expression, cloning, genetic manipulation |
| Quantity | 1 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?