missing translation for 'onlineSavingsMsg'
Learn More

L-Carnitine, 98+%

Product Code. 11458247
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458247 10 g 10g
11468247 50 g 50g
11478247 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458247 Supplier Thermo Scientific Alfa Aesar Supplier No. A17618.09
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 541-15-1 | C7H15NO3 | 161.201 g/mol

L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.

Solubility
Soluble in water, pyridine, dimethyl sulfoxide, methanol and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-15-1
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.201
MDL Number MFCD00038747
InChI Key PHIQHXFUZVPYII-ZCFIWIBFSA-N
Synonym l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine
PubChem CID 10917
ChEBI CHEBI:16347
IUPAC Name (3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
SMILES C[N+](C)(C)CC(CC(=O)[O-])O

Specifications

Melting Point ∼190°C (decomposition)
Quantity 10 g
Beilstein 4292315
Sensitivity Hygroscopic
Merck Index 14,1849
Solubility Information Soluble in water,pyridine,dimethyl sulfoxide,methanol and ethanol.
Formula Weight 161.2
Percent Purity ≥99%
Chemical Name or Material L-Carnitine
compliance-icons
Product Identifier
  • L-Carnitine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (R)-(3-Carboxy-2-hydroxypropyl)trimethyla mmonium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.