missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Citrulline, 99%

Product Code. p-3736034 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10550181

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 372-75-8 | C6H13N3O3 | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-75-8
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.19
MDL Number MFCD00064397
InChI Key RHGKLRLOHDJJDR-UHFFFAOYNA-N
Synonym l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
PubChem CID 9750
ChEBI CHEBI:16349
IUPAC Name (2S)-2-amino-5-(carbamoylamino)pentanoic acid
SMILES NC(CCCNC(N)=O)C(O)=O

Specifications

Melting Point 214°C
Color White
Ignition Residue 0.1% max.
Loss on Drying 0.3% max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NCONH(CH2)3CH(NH2)CO2H
Quantity 25 g
Merck Index 15, 2330
Specific Rotation + 25.5
Solubility Information Solubility in water: 200g/L (20°C). Other solubilities: insoluble in methanol and ethanol
Transmittance 95.0% min. (c=10, H2O, 430nm) 1cm cell
Formula Weight 175.19
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Citrulline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.