missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Cysteine, Cell Culture Reagent

Product Code. 15455929
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15455929

Brand: Thermo Scientific Chemicals J63745.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52-90-4 | C3H7NO2S | 121.154 g/mol

Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement. l-cysteine to media protects against or reverses abnormal growth & malignant transformation in aged controls. L-Cysteine is a food additive permitted for direct addition to food for human consumption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement. l-cysteine to media protects against or reverses abnormal growth & malignant transformation in aged controls. L-Cysteine is a food additive permitted for direct addition to food for human consumption.

Solubility
Soluble in water (10 mg/ml), 1N HCl (50 mg/ml), and ethanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
MDL Number MFCD00064306
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
PubChem CID 5862
ChEBI CHEBI:17561
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)S

Specifications

Melting Point ∽220°C (decomposition)
Color White
Beilstein 1721408
Merck Index 14,2781
Solubility Information Soluble in water (10mg/ml),1N HCl (50mg/ml),and ethanol.
Quantity 100 g
Formula Weight 121.16
Optical Rotation +7.6° (c=5 in 5N HCl)
Sensitivity Air Sensitive
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Cysteine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.