missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Cysteine HCl

Product Code. 30018208
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018208 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018208 Supplier Melford Supplier No. C81020100.0
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-Cysteine Hydrochloride Monohydrate is a compound that is a derivative of the amino acid cysteine and is commonly used as a precursor in the synthesis of proteins and peptides.

L-Cysteine Hydrochloride Monohydrate is a compound that is a derivative of the amino acid cysteine and is commonly used as a precursor in the synthesis of proteins and peptides.

It is often utilized in cell culture media to support the growth and viability of cells. L-Cysteine is an essential component for the synthesis of glutathione, an important antioxidant in cells.

It is employed in biotechnological processes for the production of recombinant proteins. It can act as a reducing agent, facilitating the proper folding and stability of proteins.

Researchers may use this chemical in various laboratory experiments and studies, given its role in protein synthesis and cellular processes.

Specifications

Chemical Name or Material L-Cysteine HCl
CAS 4-6-7048
Molecular Formula C3H7NO2S-HCl-H2O
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.