missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Cysteine hydrochloride monohydrate, 99%

Product Code. 11458563
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458563 100 g 100g
11448563 25 g 25g
11468563 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458563 Supplier Thermo Scientific Chemicals Supplier No. A10389.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7048-04-6 | C3H10ClNO3S | 175.63 g/mol

L-Cysteine hydrochloride monohydrate is used as a precursor in food, pharmaceutical and personal care products. It is actively involved in the production of flavors such as meat flavor obtained by reacting with sugars. It plays an important role as a key raw material for the manufacture of N-acetyl-L-cysteine, S-carboxymethyl-L-cysteine and L-cysteine base. It is involved in the treatment of liver disease as an antioxidant and antidote. Being an essential amino acid, it used as animal feed especially for sheep, which is responsible the growth of wool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Cysteine hydrochloride monohydrate is used as a precursor in food, pharmaceutical and personal care products. It is actively involved in the production of flavors such as meat flavor obtained by reacting with sugars. It plays an important role as a key raw material for the manufacture of N-acetyl-L-cysteine, S-carboxymethyl-L-cysteine and L-cysteine base. It is involved in the treatment of liver disease as an antioxidant and antidote. Being an essential amino acid, it used as animal feed especially for sheep, which is responsible the growth of wool.

Solubility
Soluble in cold water.

Notes
Air and light sensitive. Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, metals and hydrogen chloride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-6-7048
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
MDL Number MFCD00065606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
PubChem CID 23462
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Melting Point ∽175°C (decomposition)
Assay Percent Range 99%
Beilstein 5158059
Solubility Information Soluble in cold water.
Quantity 100 g
Formula Weight 175.64 (157.62 Anhydrous)
Sensitivity Air Sensitive
Chemical Name or Material L-Cysteine hydrochloride monohydrate
compliance-icons
Product Identifier
  • L-Cysteine hydrochloride monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: L-(+)-Cysteine hydrochloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.