missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine Hydrochloride, Monohydrate, Certified AR for Analysis, Fisher Chemical

Product Code. 10499740 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10499740 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10499740 Supplier Fisher Chemical Supplier No. C/9152/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 175°C
CAS 4-6-7048
pH 1 to 2
Color White
Molecular Formula C3H10ClNO3S
Packaging Plastic Powder Jar
MDL Number 65606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
Molecular Weight (g/mol) 175.63
Quantity 100 g
Formula Weight 175.64g/mol
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.