missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine Hydrochloride Monohydrate, MP Biomedicals

Product Code. 11473400 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473400 25 g 25g
11493400 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473400 Supplier MP Biomedicals Supplier No. 02101446.3
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • L-Cysteine hydrochloride monohydrate has been used in a protocol for the separation of dorsal root ganglion neurons (DRGs)
  • It has also been used in a study to examine its effect as an inhibitor in preventing enzymatic browning in apples

Chemical Identifiers

Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
MDL Number MFCD00065606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N

Specifications

CAS 4-6-7048
Melting Point 175°C (literature)
Linear Formula C3H7NO2S·HCl· H2O
Quantity 25 g
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
Solubility Information Freely soluble in water (50mg/mL).
SMILES C(C(C(=O)O)N)S.O.Cl
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
PubChem CID 23462
Formula Weight 175.64
Physical Form Crystals
Chemical Name or Material L-CYSTEINE HYDROCHLORIDE MONOHYDRATE
Show More Show Less
compliance-icons
Product Identifier
  • L-Cysteine Hydrochloride Monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: L-Cysteine Hydrochloride Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.