missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine, TRC

Product Code. 30009607
Change view
Click to view available options
Quantity:
10 mg
2.5 g
10 g
Unit Size:
10g
10mg
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009607 10 mg 10mg
30004475 2.5 g 2.5g
17960375 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009607 Supplier Toronto Research Chemicals Supplier No. TRCC99500010MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3 H7 N O2 S
Molecular Weight (g/mol) 121.16
Synonym L-Cysteine, Acetylcysteine Imp. B (EP)
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES N[C@@H](CS)C(=O)O

Specifications

InChI Formula InChI=1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m0/s1
Quantity 10 mg
Formula Weight 121.0197
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Cysteine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.