missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, 98.5 to 101.5% (Dry Basis), Thermo Scientific™

Product Code. 15835118 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
50g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15835118 50 g Plastic Bottle 50g
15805278 1 kg Plastic Bottle 1kg
15815278 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15835118 Supplier Thermo Scientific Supplier No. J14050.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
MDL Number MFCD00064228
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
PubChem CID 67678
ChEBI CHEBI:16283
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Melting Point >240°C (decomposition)
Packaging Plastic Bottle
Beilstein 1728094
Density 1.68
Quantity 50 g
Formula Weight 240.3
Percent Purity 98.5 to 101.5%
Specific Gravity 1.68
Physical Form Crystalline Powder
Chemical Name or Material L-Cystine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.