missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, Certified AR for Analysis, Fisher Chemical™

Product Code. 10010070 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10010070 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10010070 Supplier Fisher Chemical Supplier No. C/9160/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 260°C
CAS 56-89-3
Color White
Molecular Formula C6H12N2O4S2
Packaging Plastic Powder Jar
MDL Number 64228
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
SMILES N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
IUPAC Name (2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
Molecular Weight (g/mol) 240.29
Quantity 25 g
Formula Weight 240.29g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.