missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, specified according to the requirements of Ph.Eur.

Product Code. p-7077266 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15289916 25 g 25g
15299916 100 g 100g
15279916 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15289916 Supplier Thermo Scientific Acros Supplier No. 406160250
Add to basket
This item is not returnable. View return policy

CAS: 56-89-3 | C6H12N2O4S2 | 240.292 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
PubChem CID 67678
ChEBI CHEBI:16283
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Melting Point 260.0°C to 261.0°C
Ammonium 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.5% max. (1 g, 105°C)
Linear Formula [-SCH2CH(NH2)CO2H]2
Specific Rotation −224.00
Sulfate 300ppm max.
Sulfated Ash 0.1% max. (1 g)
Solubility Information Solubility in water: 0.112g/l(25°C), 24mg/100mL(50°C). Other solubilities: 52 mg/100mL water (75°C),114 mg/100mL water(100°C), soluble in aq. solutions <ph 2, soluble in mineral acids, alkalis, ammonium hydrox, ide,, insoluble in ethanol, diethyl ether, b
Specific Rotation Condition −224.00 (20.00°C c=2;1 M HCL dry.sub.)
Quantity 25 g
Formula Weight 240.3
Chemical Name or Material L-Cystine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.