missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, TRC

Product Code. 17908774
Change view
Click to view available options
Quantity:
10 mg
10 g
25 g
Unit Size:
10g
10mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17908774 10 g 10g
17967254 10 mg 10mg
17997904 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17908774 Supplier Toronto Research Chemicals Supplier No. TRCC99786010G
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6 H12 N2 O4 S2
Molecular Weight (g/mol) 240.3
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES N[C@@H](CSSC[C@H](N)C(=O)O)C(=O)O

Specifications

Synonym L-Cystine, Acetylcysteine Imp. A (EP), Cystine, L- (8CI), (-)-Cystine, 3,3'-Dithiobis(2-aminopropanoic acid), Bis(β-amino-β-carboxyethyl) disulfide, Cystine, Cystine acid, Dicysteine, L-(-)-Cystine, L-Alanine, 3,3'-dithiobis-, L-Cysteine disulfide, L-Cystin, NSC 13203, Oxidized L-cysteine, Propanoic acid, 3,3'-dithiobis[2-amino-, [R-(R*,R*)]-, [R-(R*,R*)]-3,3'-Dithiobis[2-aminopropanoic acid], l-Cystine, β,β'-Diamino-β,β'-dicarboxydiethyl disulfide, β,β'-Dithiodialanine
InChI Formula InChI=1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1
Quantity 10 g
Formula Weight 240.0238
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Cystine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.