missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine (White Crystals or Cryst. Powder), Fisher BioReagents

Product Code. 10204683
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10204683 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10204683 Supplier Fisher BioReagents Supplier No. BP377100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 260°C
Ammonium 0.02% max.
CAS 56-89-3
Arsenic (As) 1ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.3% max.
Other Amino Acids Chromatographically not detectable
Molecular Formula C6H12N2O4S2
Packaging Glass Bottle
Specific Rotation -215 to -225° (+ or -)
Merck Index 15, 2779
State of Solution (%T) 98.0% min.
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
Alpha Vector Cystine
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
Molecular Weight (g/mol) 240.292
Quantity 100 g
PubChem CID 67678
ChEBI CHEBI:16283
Formula Weight 240.29
Percent Purity 98.5 to 101.0%
Physical Form Powder
Chemical Name or Material L-Cystine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.