missing translation for 'onlineSavingsMsg'
Learn More

L-Dierucoyl Lecithin, TRC

Product Code. 30377666
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377666 100 mg
30376670 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30377666 Supplier Toronto Research Chemicals Supplier No. TRCD441945100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51779-95-4
Molecular Formula C52H100NO8P
Molecular Weight (g/mol) 898.33
Synonym (7R,22Z)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[[(13Z)-1-oxo-13-docosen-1-yl]oxy]-3,5,9-trioxa-4-phosphahentriacont-22-en-1-aminium 4-Oxide Inner Salt, [R-(Z,Z)]-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxo-13-docosenyl)oxy]-3,5,9-trioxa-4-phosphahentriacont-22-en-1-aminium Inner Salt 4-Oxide, (7R,22Z)- 4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[[(13Z)-1-oxo-13-docosenyl]oxy]-3,5,9-trioxa-4-phosphahentriacont-22-en-1-aminium Inner Salt 4-Oxide, 1,2-Dierucoyl-sn-glycero-3-phosphocholine, 1,2-Dierucoyl-sn-glycerol-3-phosphorylcholine, DEPC, Dierucoyl-L-alpha-glycerophosphorylcholine, Dierucoyllecithin
IUPAC Name [(2R)-2,3-bis[[(Z)-docos-13-enoyl]oxy]propyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCC\C=C/CCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C52H100NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-51(54)58-48-50(49-60-62(56,57)59-47-46-53(3,4)5)61-52(55)45-43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,50H,6-19,24-49H2,1-5H3/b22-20-,23-21-/t50-/m1/s1
Formula Weight 897.7187
Chemical Name or Material L-Dierucoyl Lecithin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.