missing translation for 'onlineSavingsMsg'
Learn More

L-Dihydroorotic acid, 98%

Product Code. 15436898
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436898 1 g 1g
15446898 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436898 Supplier Thermo Scientific Alfa Aesar Supplier No. H60429.03
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5988-19-2 | C5H6N2O4 | 158.113 g/mol

L-Dihydroorotic acid a metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Dihydroorotic acid a metabolite.

Solubility
Soluble in water (partly), and dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5988-19-2
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.113
MDL Number MFCD00085339
InChI Key UFIVEPVSAGBUSI-REOHCLBHSA-N
Synonym l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate
PubChem CID 439216
ChEBI CHEBI:17025
IUPAC Name (4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
SMILES C1C(NC(=O)NC1=O)C(=O)O

Specifications

Melting Point 254°C to 255°C (decomposition)
Quantity 1 g
Solubility Information Soluble in water (partly),and dimethyl formamide.
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material L-Dihydroorotic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.